missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-methylisoxazole, 98%, Thermo Scientific™

Product Code. 10033630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10033630 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10033630 Supplier Thermo Scientific Acros Supplier No. 305870010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 14678-02-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 81.0°C to 84.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H6N2O
MDL Number MFCD00003151
Quantity 1 g
Synonym 5-amino-3-methylisoxazole, 3-methylisoxazol-5-amine, 5-isoxazolamine, 3-methyl, 3-methyl-5-isoxazolamine, unii-0g66r1h4rw, 3-methyl-isoxazol-5-ylamine, 3-methylisoxazole-5-ylamine, 3-methylisoxazole-5-amine, acmc-209yk8, 3-methylisooxazole-5-amine
InChI Key FNXYWHTZDAVRTB-UHFFFAOYSA-N
SMILES CC1=NOC(=C1)N
IUPAC Name 3-methyl-1,2-oxazol-5-amine
Molecular Weight (g/mol) 98.105
PubChem CID 84590
Formula Weight 98.1
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 5-Amino-3-methylisoxazole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.