missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-3-methyl-1-phenyl-1H-pyrazole, ≥99%, Thermo Scientific Chemicals

Product Code. p-7055109
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 15420389

Brand: Thermo Scientific Alfa Aesar H63605.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-18-6
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
MDL Number MFCD00020727
InChI Key FMKMKBLHMONXJM-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-1h-pyrazol-5-amine, 5-amino-3-methyl-1-phenylpyrazole, 1h-pyrazol-5-amine, 3-methyl-1-phenyl, 1-phenyl-3-methyl-5-aminopyrazole, 3-methyl-1-phenylpyrazol-5-ylamine, 5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 1-phenyl-3-methyl-5-amino pyrazole, unii-x6087geb2p, pyrazole, 5-amino-3-methyl-1-phenyl, 1h-pyrazole-5-amine, 3-methyl-1-phenyl
PubChem CID 70801
IUPAC Name 5-methyl-2-phenylpyrazol-3-amine
SMILES CC1=NN(C(=C1)N)C2=CC=CC=C2

Specifications

Melting Point 114°C to 117°C
Color Brown to Yellow
Quantity 5 g
Beilstein 140666
Formula Weight 173.21
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 5-Amino-3-methyl-1-phenyl-1H-pyrazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.