missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzothiazole, 99%

Product Code. 11495667
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495667 2 g 2g
11405677 10 g 10g
11415677 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495667 Supplier Thermo Scientific Alfa Aesar Supplier No. B23631.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13382-43-9 | C8H8N2S | 164.23 g/mol

5-Amino-2-methylbenzothiazole is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-methylbenzothiazole is used as pharmaceutical intermediates.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13382-43-9
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00226291
InChI Key GPWQHYMVUZYWIK-UHFFFAOYSA-N
Synonym 2-methylbenzo d thiazol-5-amine, 5-amino-2-methylbenzothiazole, 5-benzothiazolamine, 2-methyl, 2-methyl-5-benzothiazolamine, 2-methyl-5-aminobenzothiazole, 2-methylbenzothiazole-5-ylamine, 2-methyl-1,3-benzothiazol-5-ylamine, acmc-209v5w, cambridge id 5107842, 2-methylbenzothiazol-5-amine
PubChem CID 36229
IUPAC Name 2-methyl-1,3-benzothiazol-5-amine
SMILES CC1=NC2=CC(N)=CC=C2S1

Specifications

Quantity 2 g
Formula Weight 164.22
Percent Purity 99%
Chemical Name or Material 5-Amino-2-methylbenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.