missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-hydroxymethylphenylboronic Acid Hydrochloride, TRC

Product Code. 30319256
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319256 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319256 Supplier Toronto Research Chemicals Supplier No. TRCA6279831G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Amino-2-hydroxymethylphenylboronic Acid Hydrochloride
CAS 117098-93-8
Molecular Formula C7H8BNO2 . HCl
Quantity 1 g
Synonym 6-Amino-1,3-dihydro-1-hydroxy-2,1-benzoxaborole Hydrochloride, 1,3-Dihydro-1-hydroxy-2,1-benzoxaborol-6-amine Hydrochloride
InChI Formula InChI=1S/C7H8BNO2.ClH/c9-6-2-1-5-4-11-8(10)7(5)3-6;/h1-3,10H,4,9H2;1H
SMILES Cl.Nc1ccc2COB(O)c2c1
IUPAC Name 1-hydroxy-3H-2,1-benzoxaborol-6-amine;hydrochloride
Molecular Weight (g/mol) 185.42
Formula Weight 185.0415
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.