missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-chloropyrimidine, 95%

Product Code. 10077809 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10077809 5 g Glass bottle 5g
10872451 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10077809 Supplier Thermo Scientific Acros Supplier No. 434320050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56621-90-0 | C4H4ClN3 | 129.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56621-90-0
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
InChI Key DZBKIOJXVOECRA-UHFFFAOYSA-N
PubChem CID 12215993
IUPAC Name 2-chloropyrimidin-5-amine
SMILES C1=C(C=NC(=N1)Cl)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 160°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Synonym 5-amino-2-chloropyrimidine, 2-chloro-pyrimidin-5-ylamine, 2-chloro-5-aminopyrimidine, 5-pyrimidinamine, 2-chloro, 2-chloropyrimidine-5-ylamine, pubchem6963, acmc-1axfh, 2-chloropyrimidin-5-ylamine, 5-pyrimidinamine,2-chloro, 5-amino-2-chloro pyrimidine
Formula Weight 129.55
Percent Purity 95%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 5-Amino-2-chloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-2-chloropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.