missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-chlorobenzoic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2-chlorobenzoic Acid |
| CAS | 89-54-3 |
| Molecular Formula | C7H6ClNO2 |
| Quantity | 10 g |
| Synonym | 2-Chloro-5-aminobenzoic Acid, 3-Amino-6-chlorobenzoic Acid, 5-Amino-2-chlorobenzoic Acid, NSC 25172 |
| InChI Formula | InChI=1S/C7H6ClNO2/c8-6-2-1-4(9)3-5(6)7(10)11/h1-3H,9H2,(H,10,11) |
| SMILES | Nc1ccc(Cl)c(c1)C(=O)O |
| IUPAC Name | 5-amino-2-chlorobenzoic acid |
| Molecular Weight (g/mol) | 171.58 |
| Formula Weight | 171.0087 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction