missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-bromobenzoic acid, 95%

Product Code. 15426848
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426848 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426848 Supplier Thermo Scientific Alfa Aesar Supplier No. H60382.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2840-02-0 | C7H6BrNO2 | 216.03 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2840-02-0
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD07368968
InChI Key FEXDUVBQBNYSQV-UHFFFAOYSA-N
Synonym 5-amino-2-bromo-benzoic acid, 4-bromo-3-carboxyaniline, benzoic acid,5-amino-2-bromo, benzoic acid, 5-amino-2-bromo, pubchem4637, acmc-209h2e, 5-amino-2-bromobenzoicacid, 2-bromo-5-aminobenzoic acid, ksc561g5h, 5-amino-2-bromo benzoic acid
PubChem CID 2769624
IUPAC Name 5-amino-2-bromobenzoic acid
SMILES NC1=CC(C(O)=O)=C(Br)C=C1

Specifications

Melting Point 171°C to 179°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 216.03
Percent Purity 95%
Chemical Name or Material 5-Amino-2-bromobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.