missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,3-dihydrobenzofuran, 97%

Product Code. 10891691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10891691 5 g 5g
10785072 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10891691 Supplier Thermo Scientific Acros Supplier No. 438030050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42933-43-7 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42933-43-7
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
InChI Key YJMADHMYUJFMQE-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-amine, 2,3-dihydrobenzo b furan-5-ylamine, 5-amino-2,3-dihydrobenzo b furan, 5-aminocoumaran, 5-amino-2,3-dihydrobenzofuran, 2,3-dihydro-5-benzofuranamine, 2,3-dihydrobenzo b furan-5-amine, pubchem16462, 2,3-dihydro-5-benzofuranamine;, 2,3-dihydrobenzofuran-5-amine;
PubChem CID 3841102
IUPAC Name 2,3-dihydro-1-benzofuran-5-amine
SMILES C1COC2=C1C=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 81.6°C to 83.4°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 135.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Amino-2, 3-dihydrobenzofuran
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-2,3-dihydrobenzofuran
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.