Learn More
5-Amino-1H-tetrazole monohydrate, 99%
Description
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.
Solubility
Soluble in water, ethanol (1.36%).
Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
Specifications
Specifications
| Melting Point | ∼203°C (decomposition) |
| pH | 3.7 |
| Quantity | 25 g |
| UN Number | UN1325 |
| Beilstein | 108872 |
| Solubility Information | Soluble in water,ethanol (1.36%). |
| Formula Weight | 103.08 (85.06 Anhydrous) |
| Percent Purity | 99% |
| Chemical Name or Material | 5-Amino-1H-tetrazole monohydrate |
Safety and Handling
- 5-Amino-1H-tetrazole monohydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contain: 2H-Tetrazol-5-amine,hydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.