missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-benzotriazole, 96%

Product Code. 11376946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376946 1 g 1g
11386946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376946 Supplier Thermo Scientific Alfa Aesar Supplier No. L06660.03
 Restricted Item
Estimated Shipment: 13-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3325-11-9 | C6H6N4 | 134.14 g/mol

5-Amino-1H-benzotriazole is used as azole building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-1H-benzotriazole is used as azole building block.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3325-11-9
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.14
MDL Number MFCD00047209
InChI Key XSFHICWNEBCMNN-UHFFFAOYSA-N
Synonym 5-aminobenzotriazole, 1h-1,2,3-benzotriazol-5-amine, 1h-benzotriazol-6-amine, 1h-benzotriazol-5-amine, 5-amino-1h-benzotriazole, 2h-benzo d 1,2,3 triazol-5-amine, 1h-benzo d 1,2,3 triazol-5-amine, benzotriazole-5-ylamine, 3h-1,2,3-benzotriazol-5-amine, 5-amino benzotriazole
PubChem CID 76844
IUPAC Name 2H-benzotriazol-5-amine
SMILES C1=CC2=NNN=C2C=C1N

Specifications

Melting Point 157°C to 162°C
Odor Odorless
Quantity 1 g
Beilstein 121262
Solubility Information Insoluble in water.
Formula Weight 134.14
Percent Purity 98%
Chemical Name or Material 5-Amino-1H-benzotriazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.