missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-benzimidazole-2-thiol, 97%, Thermo Scientific™

Product Code. 10250173
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10250173 10 g 10g
10361463 1 g 1g
10585973 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10250173 Supplier Thermo Scientific Maybridge Supplier No. AC34641EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2818-66-8
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.21
MDL Number MFCD00022671
InChI Key BXDMTLVCACMNJO-UHFFFAOYSA-N
Synonym 5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol
PubChem CID 2734002
SMILES NC1=CC=C2NC(=S)NC2=C1

Specifications

Assay Percent Range 97%
Quantity 10 g
IUPAC Name 5-amino-1,3-dihydrobenzimidazole-2-thione
Formula Weight 165.22
Chemical Name or Material 5-Amino-1H-benzimidazole-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.