missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-pentanol, 50 wt.% aqueous solution

Product Code. 10280103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
800 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
800mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10280103 800 mL Glass bottle 800mL
10462815 25 mL Glass bottle 25mL
10391253 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10280103 Supplier Thermo Scientific Acros Supplier No. 387918000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2508-29-4 | C5H13NO | 103.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2508-29-4 , 7732-18-5
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.16
MDL Number MFCD00008237
InChI Key LQGKDMHENBFVRC-UHFFFAOYSA-N
Synonym 5-amino-1-pentanol, 5-aminopentanol, 1-pentanol, 5-amino, pentanol, 5-amino, 5-amino-1-pentanol in water, 5-amino pentanol, 5-amino-l-pentanol, 5-aminopentanol-1, 5-hydroxypentylamine, 1-amino-5-pentanol
PubChem CID 75634
IUPAC Name 5-aminopentan-1-ol
SMILES C(CCN)CCO

Specifications

CAS Min % 48.0
CAS Max % 52.0
Density 0.9870g/mL
Flash Point >100°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula OH(CH2)5NH2
Quantity 800 mL
Specific Gravity 0.987
Solubility Information Solubility in water: miscible.
Formula Weight 103.16
Percent Purity 48.0 to 52.0%
Chemical Name or Material 5-Amino-1-pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Amino-1-pentanol, 50 wt.% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.