missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1-naphthalenesulfonic Acid, 90%, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 5-Amino-1-naphthalenesulfonic Acid |
| CAS | 84-89-9 |
| Molecular Formula | C10H9NO3S |
| Quantity | 25 g |
| Synonym | 1-Amino-5-naphthalenesulfonic Acid, 1-Amino-5-sulfonaphthalene, 1-Naphthylamine-5-sulfonic Acid, 5-Naphthylamine-1-sulfonic Acid, 5-Sulfo-1-naphthylamine, Laurent Acid, NSC 28691, NSC 7553 |
| InChI Formula | InChI=1S/C10H9NO3S/c11-9-5-1-4-8-7(9)3-2-6-10(8)15(12,13)14/h1-6H,11H2,(H,12,13,14) |
| SMILES | Nc1cccc2c(cccc12)S(=O)(=O)O |
| IUPAC Name | 5-aminonaphthalene-1-sulfonic acid |
| Molecular Weight (g/mol) | 223.25 |
| Formula Weight | 223.0303 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction