missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-1-methyl-1H-indazole, 97%, Thermo Scientific™
£69.90 - £271.00
Chemical Identifiers
CAS | 50593-24-3 |
---|---|
Molecular Formula | C8H9N3 |
Molecular Weight (g/mol) | 147.18 |
MDL Number | MFCD03305455 |
InChI Key | PYOFNPHTKBSXOM-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-indazol-5-amine, 5-amino-1-methyl-1h-indazole, 1-methyl-1h-indazol-5-ylamine, 1h-indazol-5-amine, 1-methyl, 5-amino-1-methylindazole, 1-methyl-1h-indazole-5-amine, pubchem20587, 1-methyl-5-aminoindazol, 1-methyl-5-indazolamine, 1-methyl-5-aminoindazole |
PubChem CID | 2768032 |
IUPAC Name | 1-methylindazol-5-amine |
SMILES | CN1C2=C(C=C(C=C2)N)C=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10309747
|
Thermo Scientific Acros
429520010 |
1 g | Glass bottle |
£69.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10760540
|
Thermo Scientific Acros
429520050 |
5 g | Glass bottle |
£271.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
50593-24-3 | |
147.18 | |
PYOFNPHTKBSXOM-UHFFFAOYSA-N | |
2768032 | |
CN1C2=C(C=C(C=C2)N)C=N1 |
C8H9N3 | |
MFCD03305455 | |
1-methyl-1h-indazol-5-amine, 5-amino-1-methyl-1h-indazole, 1-methyl-1h-indazol-5-ylamine, 1h-indazol-5-amine, 1-methyl, 5-amino-1-methylindazole, 1-methyl-1h-indazole-5-amine, pubchem20587, 1-methyl-5-aminoindazol, 1-methyl-5-indazolamine, 1-methyl-5-aminoindazole | |
1-methylindazol-5-amine |
Specifications
50593-24-3 | |
100.0 | |
Brown | |
97% min. (HPLC) | |
MFCD03305455 | |
PYOFNPHTKBSXOM-UHFFFAOYSA-N | |
1-methylindazol-5-amine | |
2768032 | |
97% | |
5-Amino-1-methyl-1H-indazole |
96.0 | |
152.0°C to 154.0°C | |
Authentic | |
C8H9N3 | |
1-methyl-1h-indazol-5-amine, 5-amino-1-methyl-1h-indazole, 1-methyl-1h-indazol-5-ylamine, 1h-indazol-5-amine, 1-methyl, 5-amino-1-methylindazole, 1-methyl-1h-indazole-5-amine, pubchem20587, 1-methyl-5-aminoindazol, 1-methyl-5-indazolamine, 1-methyl-5-aminoindazole | |
CN1C2=C(C=C(C=C2)N)C=N1 | |
147.18 | |
147.18 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Amino-1-methyl-1H-indazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only