missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylvaleric acid, 96%

Product Code. p-7022174
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324679 5 g 5g
11482557 10 g 10g
11492557 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324679 Supplier Thermo Scientific Alfa Aesar Supplier No. A14810.06
Add to basket
This item is not returnable. View return policy

CAS: 3128-07-2 | C7H12O3 | 144.17 g/mol

5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Acetylvaleric acid, is used as a reagent to synthesize new penicillins containing keto acids as side chains. It is also used to study the various metabolic pathways of 4-hydroxypentanoate and Levulinate. It is also used as an Pharmaceutical intermediate.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3128-07-2
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00051479
InChI Key IZOQMUVIDMLRDC-UHFFFAOYSA-N
Synonym 5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
PubChem CID 98810
IUPAC Name 6-oxoheptanoic acid
SMILES CC(=O)CCCCC(=O)O

Specifications

Melting Point 34°C to 36°C
Boiling Point 138°C to 140°C (1.5 mmHg)
Refractive Index 1.449
Quantity 5 g
UN Number UN3261
Beilstein 1362931
Sensitivity Hygroscopic
Solubility Information Soluble in water
Formula Weight 144.17
Percent Purity 96%
Chemical Name or Material 5-Acetylvaleric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.