missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylvaleric acid, 90+%, technical

Product Code. 13478668 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13478668 5 g 5g
13488668 25 g 25g
13498668 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13478668 Supplier Thermo Scientific Acros Supplier No. 449530050
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3128-07-2 | C7H10O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3128-07-2
Molecular Formula C7H10O3
Molecular Weight (g/mol) 144.17
InChI Key IZOQMUVIDMLRDC-UHFFFAOYSA-N
Synonym 5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
PubChem CID 98810
IUPAC Name 6-oxoheptanoic acid
SMILES CC(=O)CCCCC(=O)O

Specifications

Melting Point 35°C to 37°C
Boiling Point 138°C to 140°C (1.5 mmHg)
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(O)(CH2)4CO2H
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 144.17
Percent Purity 90+%
Grade Technical
Chemical Name or Material 5-Acetylvaleric acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Acetylvaleric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.