missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylthiophene-2-carbonitrile, 97%, Thermo Scientific™

Product Code. 10236004
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236004 25 g Amber Glass Bottle 25g
10612665 5 g Amber Glass Bottle 5g
10556423 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10236004 Supplier Thermo Scientific Maybridge Supplier No. TL00354EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 88653-55-8
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
InChI Key VSHPLUBHIUFLES-UHFFFAOYSA-N
Synonym 2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ci
PubChem CID 2747565
IUPAC Name 5-acetylthiophene-2-carbonitrile
SMILES CC(=O)C1=CC=C(S1)C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material 5-Acetylthiophene-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.