missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylthiophene-2-boronic acid, 98%

Product Code. 11348216
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11348216

Brand: Thermo Scientific Alfa Aesar L15221.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 206551-43-1 | C6H7BO3S | 169.989 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206551-43-1
Molecular Formula C6H7BO3S
Molecular Weight (g/mol) 169.989
MDL Number MFCD01075681
InChI Key DCNMATSPQKWETQ-UHFFFAOYSA-N
Synonym 5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
PubChem CID 4143377
IUPAC Name (5-acetylthiophen-2-yl)boronic acid
SMILES B(C1=CC=C(S1)C(=O)C)(O)O

Specifications

Melting Point 142°C to 148°C
Quantity 25 g
Beilstein 8681302
Formula Weight 170
Percent Purity 98%
Chemical Name or Material 5-Acetylthiophene-2-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.