missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Acetylsalicylic Acid Methyl Ester, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 5-Acetylsalicylic Acid Methyl Ester |
| CAS | 16475-90-4 |
| Molecular Formula | C10H10O4 |
| Quantity | 1 g |
| Synonym | 5-Acetyl-2-hydroxy-benzoic Acid Methyl Ester, Methyl 2-Hydroxy-5-acetylbenzoate, Methyl 3-Acetyl-6-hydroxybenzoate, Methyl 5-Acetyl-2-hydroxybenzoate, Methyl 5-Acetylsalicylate, NSC 67867 |
| InChI Formula | InChI=1S/C10H10O4/c1-6(11)7-3-4-9(12)8(5-7)10(13)14-2/h3-5,12H,1-2H3 |
| SMILES | COC(=O)c1cc(ccc1O)C(=O)C |
| IUPAC Name | methyl 5-acetyl-2-hydroxybenzoate |
| Molecular Weight (g/mol) | 194.18 |
| Formula Weight | 194.0579 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction