missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylsalicylic acid, 98+%

Product Code. 11499133
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499133 100 g 100g
11479133 5 g 5g
11489133 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499133 Supplier Thermo Scientific Alfa Aesar Supplier No. A11663.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13110-96-8 | C9H8O4 | 180.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13110-96-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013978
InChI Key NZRDKNBIPVLNHA-UHFFFAOYSA-N
PubChem CID 83151
IUPAC Name 5-acetyl-2-hydroxybenzoic acid
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 214°C to 216°C
Flash Point 250°C (482°F)
Quantity 100 g
Beilstein 2096968
Sensitivity Moisture sensitive
Synonym 5-acetylsalicylic acid, 3-acetyl-6-hydroxybenzoic acid, 5-acetyl-2-hydroxy-benzoic acid, benzoic acid, 5-acetyl-2-hydroxy, 5-acetyl salicylic acid, acmc-1boxk, 5-acetylsalicylicacid, 5-acetyl-2-hydroxybenzoic acid, benzoic acid,5-acetyl-2-hydroxy
Formula Weight 180.15
Percent Purity ≥98%
Chemical Name or Material 5-Acetylsalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.