missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylsalicylamide, 98%, Thermo Scientific™

Product Code. 11403857
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403857 5 g 5g
11413857 25 g 25g
11423857 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403857 Supplier Thermo Scientific Alfa Aesar Supplier No. A18303.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40187-51-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00049222
InChI Key LWAQTCWTCCNHJR-UHFFFAOYSA-N
Synonym 5-acetylsalicylamide, benzamide, 5-acetyl-2-hydroxy, 5-acetyl-2-hydroxy-benzamide, 5-acetyl salicylamide, acmc-1alb7, dsstox_cid_31525, dsstox_rid_97410, dsstox_gsid_57736, 4-10-00-03635 beilstein handbook reference
PubChem CID 198212
IUPAC Name 5-acetyl-2-hydroxybenzamide
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)N

Specifications

Melting Point 220°C to 222°C
Quantity 5 g
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 5-Acetylsalicylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.