missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-hydroxybenzamide, 97%, Thermo Scientific™

Product Code. 10535754
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10535754 50 g 50g
10062114 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10535754 Supplier Thermo Scientific Maybridge Supplier No. SB01472EE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 40187-51-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
InChI Key LWAQTCWTCCNHJR-UHFFFAOYSA-N
Synonym 5-acetylsalicylamide, benzamide, 5-acetyl-2-hydroxy, 5-acetyl-2-hydroxy-benzamide, 5-acetyl salicylamide, acmc-1alb7, dsstox_cid_31525, dsstox_rid_97410, dsstox_gsid_57736, 4-10-00-03635 beilstein handbook reference
PubChem CID 198212
IUPAC Name 5-acetyl-2-hydroxybenzamide
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)N

Specifications

Assay Percent Range 97%
Quantity 50 g
Formula Weight 179.17
Chemical Name or Material 5-Acetyl-2-hydroxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.