missing translation for 'onlineSavingsMsg'
Learn More

5-Acetoxymethyl-2-furaldehyde, 97%

Product Code. 10752201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10752201 2 g 2g
10205500 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752201 Supplier Thermo Scientific Acros Supplier No. 163400020
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10551-58-3 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10551-58-3
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00003233
InChI Key QAVITTVTXPZTSE-UHFFFAOYSA-N
Synonym 5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
PubChem CID 66349
IUPAC Name (5-formylfuran-2-yl)methyl acetate
SMILES CC(=O)OCC1=CC=C(O1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 52°C to 56°C
Color Brown
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 2 g
Beilstein 18, 15
Formula Weight 168.15
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 5-Acetoxymethyl-2-furaldehyde, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.