missing translation for 'onlineSavingsMsg'
Learn More

5-Acetoxymethyl-2-furaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-48389
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
10551-58-3
C8H8O4
168.15
MFCD00003233
QAVITTVTXPZTSE-UHFFFAOYSA-N
5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
66349
(5-formylfuran-2-yl)methyl acetate
CC(=O)OCC1=CC=C(O1)C=O
This item is not returnable. View return policy
10551-58-3
C8H8O4
168.15
MFCD00003233
QAVITTVTXPZTSE-UHFFFAOYSA-N
5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
66349
(5-formylfuran-2-yl)methyl acetate
CC(=O)OCC1=CC=C(O1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 52°C to 56°C
Color Brown
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 2 g
Beilstein 18, 15
Formula Weight 168.15
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 5-Acetoxymethyl-2-furaldehyde, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'