missing translation for 'onlineSavingsMsg'
Learn More

5-Acetamido-2-nitrobenzoic acid, 98%, Thermo Scientific™

Product Code. 11418957
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418957 10 g 10g
11428957 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418957 Supplier Thermo Scientific Alfa Aesar Supplier No. B20568.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4368-83-6
Molecular Formula C9H8N2O5
Molecular Weight (g/mol) 224.172
MDL Number MFCD00035889
InChI Key ZSHFMOUMOUOGKI-UHFFFAOYSA-N
Synonym 5-acetamido-2-nitrobenzoic acid, 5-acetylamino-2-nitrobenzoic acid, benzoic acid, 5-acetylamino-2-nitro, 2-nitro-5-acetylaminobenzoic acid, 5-acetamido-2-nitrobenzoicacid, pubchem13220, acmc-209jv7, 5-acetamido-2-nitrobenzoic
PubChem CID 78076
ChEBI CHEBI:86569
SMILES CC(=O)NC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Quantity 10 g
Formula Weight 224.17
Percent Purity 98%
Chemical Name or Material 5-Acetamido-2-nitrobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.