missing translation for 'onlineSavingsMsg'
Learn More

5,8-Epoxy-9-cis Retinoic Acid, TRC

Product Code. 30384148
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384148 1 mg 1mg
30377032 10 mg 10mg
30375464 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384148 Supplier Toronto Research Chemicals Supplier No. TRCE5898251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,8-Epoxy-9-cis Retinoic Acid
Analyte or Component Names 5,8-Epoxy-9-cis Retinoic Acid
Shipping Condition Room Temperature
Molecular Formula C20 H28 O3
InChI Formula InChI=1S/C20H28O3/c1-14(12-18(21)22)8-6-9-15(2)16-13-17-19(3,4)10-7-11-20(17,5)23-16/h6,8-9,12-13,16H,7,10-11H2,1-5H3,(H,21,22)/b8-6-,14-12-,15-9-
Solution Type Neat
SMILES C\C(=C\C(=O)O)\C=C/C=C(/C)\C1OC2(C)CCCC(C)(C)C2=C1
IUPAC Name (2Z,4Z,6Z)-7-(4,4,7a-trimethyl-2,5,6,7-tetrahydro-1-benzofuran-2-yl)-3-methylocta-2,4,6-trienoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 316.435
Quantity 1 mg
Formula Weight 316.204
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.