missing translation for 'onlineSavingsMsg'
Learn More

5,8,11,14-Eicosatetraynoic acid

Product Code. 15474189
Change view
Click to view available options
Quantity:
20 mg
100 mg
Unit Size:
100mg
20mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474189 100 mg 100mg
15464189 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474189 Supplier Thermo Scientific Alfa Aesar Supplier No. J61624.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor | CAS: 1191-85-1 | C20H24O2 | 296.41 g/mol

A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-85-1
Molecular Formula C20H24O2
Molecular Weight (g/mol) 296.41
MDL Number MFCD00036967
InChI Key MGLDCXPLYOWQRP-UHFFFAOYSA-N
Synonym etya, 5,8,11,14-eicosatetraynoic acid, octadehydroarachidonic acid, eicosatetranoic acid, opera_id_402, spectrum5_001952, d0g9ip, bml2-f04, 5,8,11,14-eicosatetraynoicacid, 5,8,11,14-icosatetraynoic acid #
PubChem CID 1780
IUPAC Name icosa-5,8,11,14-tetraynoic acid
SMILES CCCCCC#CCC#CCC#CCC#CCCCC(=O)O

Specifications

Melting Point 81°C to 82°C
Color White
Linear Formula CH3(CH2)4(C≡CCH2)4CH2CH2CO2H
Quantity 100 mg
Beilstein 1798411
Formula Weight 296.4
Physical Form Solid
Chemical Name or Material 5,8,11,14-Eicosatetraynoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.