missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dihydroxy-4-methyl-2H-chromen-2-one Monohydrate, TRC

Product Code. 30382557
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382557 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382557 Supplier Toronto Research Chemicals Supplier No. TRCD453263100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,7-Dihydroxy-4-methyl-2H-chromen-2-one Monohydrate
CAS 189251-57-8
Molecular Formula C10H8O4 • H2O
Quantity 100 mg
InChI Formula InChI=1S/C10H8O4/c1-5-2-9(13)14-8-4-6(11)3-7(12)10(5)8/h2-4,11-12H,1H3
SMILES CC1=CC(=O)OC2=CC(=CC(=C12)O)O
Molecular Weight (g/mol) 210.181802
Formula Weight 210.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.