missing translation for 'onlineSavingsMsg'
Learn More

5,7-Difluoro-1-indanone, 97%

Product Code. p-7035794
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430438 1 g 1g
15440438 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430438 Supplier Thermo Scientific Alfa Aesar Supplier No. H54002.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84315-25-3 | C9H6F2O | 168.14 g/mol

5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Difluoro-1-indanone is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.081 g/L 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84315-25-3
Molecular Formula C9H6F2O
Molecular Weight (g/mol) 168.14
MDL Number MFCD09908161
InChI Key XWZGNWCKXLECBO-UHFFFAOYSA-N
Synonym 5,7-difluoro-1-indanone, 5,7-difluoro-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5,7-difluoro-2,3-dihydro, 5,7-difluoroindan-1-one, 1h-inden-1-one,5,7-difluoro-2,3-dihydro, acmc-20a83k, 5,7-difluoro-2,3-dihydro-1h-indene-1-one
PubChem CID 11789595
IUPAC Name 5,7-difluoro-2,3-dihydroinden-1-one
SMILES FC1=CC(F)=C2C(=O)CCC2=C1

Specifications

Melting Point 81°C
Quantity 1 g
Solubility Information Sparingly soluble in water 0.081g/L 25°C.
Formula Weight 168.14
Percent Purity 97%
Chemical Name or Material 5,7-Difluoro-1-indanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.