missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichlorokynurenic acid sodium salt, Thermo Scientific™

Product Code. 15414159
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414159 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15414159 Supplier Thermo Scientific Alfa Aesar Supplier No. J61567.MF

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent NMDA receptor glycine site antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5,7-Dichlorokynurenic acid sodium salt is a potent NMDA receptor glycine site antagonist. It acts as a chemical and organic intermediate.

Solubility
Soluble in DMSO. Insoluble in water

Notes
May be stored at 4°C. For long-term storage, aliquot and store at 4°C. Do not freeze. For maximum recovery of product, centrifuge the original vial prior to removing the cap. Further dilutions can be made in assay buffer. Protect from light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184986-70-6
Molecular Formula C10H4Cl2NNaO3
Molecular Weight (g/mol) 280.036
MDL Number MFCD00674925
InChI Key VPRPMJHKWHCUFW-UHFFFAOYSA-M
Synonym 5,7-dichlorokynurenic acid sodium salt, sodium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, potassium 5,7-dichloro-4-hydroxyquinoline-2-carboxylate, 5,7-dichloro-4-hydroxyquinoline-2-carboxylic acid sodium salt
PubChem CID 44134672
IUPAC Name sodium;5,7-dichloro-4-oxo-1H-quinoline-2-carboxylate
SMILES C1=C(C=C2C(=C1Cl)C(=O)C=C(N2)C(=O)[O-])Cl.[Na+]

Specifications

Color Yellow
Quantity 50 mg
Solubility Information Soluble in DMSO; Insoluble in water
Formula Weight 280.04
Physical Form Solid
Chemical Name or Material 5,7-Dichlorokynurenic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.