missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5,7-Dichloro-8-hydroxyquinoline, 99%

Product Code. 11450268
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450268 25 g 25g
11460268 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450268 Supplier Alfa Aesar™ Supplier No. B23809.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 773-76-2
Molecular Formula C9H5Cl2NO
Molecular Weight (g/mol) 214.045
MDL Number MFCD00006786
InChI Key WDFKMLRRRCGAKS-UHFFFAOYSA-N
Synonym chloroxine, 5,7-dichloro-8-hydroxyquinoline, chlorquinol, capitrol, dichloroxin, quixalin, chloroxyquinoline, dichloroquinolinol, dikhloroskin, 5,7-dichloro-8-quinolinol
PubChem CID 2722
ChEBI CHEBI:59477
IUPAC Name 5,7-dichloroquinolin-8-ol
SMILES C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1

Specifications

Melting Point 180°C to 182°C
Quantity 25 g
Beilstein 153606
Merck Index 14,2175
Formula Weight 214.05
Percent Purity 99%
Chemical Name or Material 5,7-Dichloro-8-hydroxyquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.