missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro-1,6-naphthyridine, 97%

Product Code. 11358431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
11358431 5 g Glass bottle 5g
11926711 1 g Glass bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 11358431

Brand: Thermo Scientific Acros 440670050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 337958-60-8 | C8H4Cl2N2 | 199.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 337958-60-8
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.04
InChI Key HNRPBMNTCYRAJD-UHFFFAOYSA-N
Synonym pubchem14653, 5,7-dichloro-1,6-napthyridine, 1,6-naphthyridine,5,7-dichloro, 1,6-naphthyridine, 5,7-dichloro, 5,7-bis chloranyl-1,6-naphthyridine
PubChem CID 12204233
IUPAC Name 5,7-dichloro-1,6-naphthyridine
SMILES C1=CC2=C(N=C(C=C2N=C1)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 138°C to 140°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 199.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5, 7-Dichloro-1, 6-naphthyridine
Show More Show Less

RUO – Research Use Only

Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.