missing translation for 'onlineSavingsMsg'
Learn More

5,6-trans-Vitamin D3, ~90%, TRC

Product Code. 18011220
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18011220 25 mg 25mg
18003810 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18011220 Supplier Toronto Research Chemicals Supplier No. TRCV67606525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22350-41-0
Molecular Formula C27 H44 O
Molecular Weight (g/mol) 384.64
Synonym (1S,3E)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-Dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylenecyclohexanol, 9,10-Secocholesta-5,7,10(19)-trien-3-ol, (3β,5E,7E)- (9CI), 9,10-Secocholesta-5,7,10(19)-trien-3β-ol, (E,E)- (8CI), 5,6-trans-Cholecalciferol, 5,6-trans-Vitamin D3, trans-Vitamin D3, Cholecalciferol impurity A, Trans-cholecalciferol
IUPAC Name (1S,3E)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C\3/C[C@@H](O)CCC3=C)\CCC[C@]12C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13+/t21-,24+,25-,26+,27-/m1/s1
Formula Weight 384.3392
Chemical Name or Material 5,6-trans-Vitamin D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.