missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethylbenzimidazole, 99+%

Product Code. 10540741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10540741 25 g Glass bottle 25g
10306750 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10540741 Supplier Thermo Scientific Acros Supplier No. 115980250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 582-60-5 | C9H10N2 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-60-5
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00005603
InChI Key LJUQGASMPRMWIW-UHFFFAOYSA-N
Synonym 5,6-dimethylbenzimidazole, dimedazol, dimedazole, dimesol, dimezol, 5,6-dimethyl-1h-benzo d imidazole, dimethylbenzimidazole, 1h-benzimidazole, 5,6-dimethyl, benzimidazole, 5,6-dimethyl, 5.6-dimethylbenzimidazole
PubChem CID 675
ChEBI CHEBI:15890
IUPAC Name 5,6-dimethyl-1H-1,3-benzodiazole
SMILES CC1=C(C)C=C2N=CNC2=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 203°C to 206°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 3258
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in diluted acids,soluble in chloroform,ether and ethanol
Formula Weight 146.19
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 5, 6-Dimethylbenzimidazole, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.