missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethyl-2,1,3-benzothiadiazole, 97%

Product Code. 11415626
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415626 1 g 1g
11425626 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415626 Supplier Thermo Scientific Alfa Aesar Supplier No. B20755.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

CAS: 1887-60-1 | C8H8N2S | 164.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1887-60-1
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.226
MDL Number MFCD00174277
InChI Key QQCMLCGOLBUDJF-UHFFFAOYSA-N
Synonym 5,6-dimethylbenzo-2,1,3-thiadiazole, 5,6-dimethylbenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazole,5,6-dimethyl, 5,6-dimethylpiazthiole, acmc-1bqjw, maybridge1_006083, 5,6-dimethylbenzo c 1,2,5-thiadiazole, 5,6-dimethyl-2,1,3-benzothiadiazole
PubChem CID 690255
IUPAC Name 5,6-dimethyl-2,1,3-benzothiadiazole
SMILES CC1=CC2=NSN=C2C=C1C

Specifikationer

Melting Point 83°C to 85°C
Quantity 1 g
Formula Weight 164.23
Percent Purity 97%
Chemical Name or Material 5,6-Dimethyl-2,1,3-benzothiadiazole

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.