missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethyl-1H-benzotriazole hydrate, 99%

Product Code. 10366840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366840 5 g Glass bottle 5g
10020850 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366840 Supplier Thermo Scientific Acros Supplier No. 116050050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4184-79-6 | C8H9N3 | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4184-79-6
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD00149344
InChI Key MVPKIPGHRNIOPT-UHFFFAOYSA-N
Synonym 5,6-dimethyl-1h-benzotriazole hydrate, 5,6-dimethyl-1,2,3-benzotriazole hydrate, 5,6-dimethyl-1h-1,2,3-benzotriazole hydrate, acmc-1aqbb, 1h-benzotriazole,5,6-dimethyl, 5,6-dimethylbenzotriazole, oxamethane, 5,6-dimethyl-2h-benzotriazole hydrate, 5,6-dimethyl-1h-benzotriazole monohydrate, 5,6-dimethyl-1h-benzo d 1,2,3 triazole hydrate
PubChem CID 16219214
IUPAC Name 5,6-dimethyl-2H-benzotriazole;hydrate
SMILES CC1=C(C)C=C2N=NNC2=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 153.0°C to 156.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Formula Weight 147.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5, 6-Dimethyl-1H-benzotriazole hydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.