missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethyl-1,2,3-benzotriazole, TRC

Product Code. 30375407
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375407 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375407 Supplier Toronto Research Chemicals Supplier No. TRCD477105100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,6-Dimethyl-1H-benzotriazole
CAS 4184-79-6
Molecular Formula C8 H8 N3
Quantity 100 mg
Synonym Benzotriazole, 5,6-dimethyl- (6CI,7CI), 5,6-Dimethyl-1H-benzotriazole, 5,6-Dimethyl-1,2,3-benzotriazole, 5,6-Dimethylbenzotriazole, NSC 62005
InChI Formula InChI=1S/C8H9N3/c1-5-3-7-8(4-6(5)2)10-11-9-7/h3-4H,1-2H3,(H,9,10,11)
SMILES Cc1cc2nn[nH]c2cc1C
IUPAC Name 5,6-dimethyl-1H-benzotriazole
Molecular Weight (g/mol) 147.18
Formula Weight 147.0796
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.