missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydrodihydroxycarbaryl, TRC

Product Code. 30387504
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387504 10 mg
30369132 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387504 Supplier Toronto Research Chemicals Supplier No. TRCD44027010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5375-49-5
Molecular Formula C12 H13 N O4
Molecular Weight (g/mol) 235.24
Synonym 1,2,5-Naphthalenetriol, 1,2-dihydro-, 5-(methylcarbamate) (9CI), Carbamic acid, methyl-, 1,2-dihydro-1,2-dihydroxy-5-naphthyl ester (7CI), Carbamic acid, methyl-, 5,6-dihydro-5,6-dihydroxy-1-naphthyl ester (8CI), 5,6-Dihydro-5,6-dihydroxy-1-naphthyl N-methylcarbamate, 5,6-Dihydro-5,6-dihydroxy-1-naphthyl methylcarbamate, 5,6-Dihydro-5,6-dihydroxycarbaryl, 5,6-Dihydro-5,6-dihydroxynaphthol methylcarbamate, 5,6-Dihydrodihydroxycarbaryl
IUPAC Name (5,6-dihydroxy-5,6-dihydronaphthalen-1-yl) N-methylcarbamate
SMILES CNC(=O)Oc1cccc2C(O)C(O)C=Cc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H13NO4/c1-13-12(16)17-10-4-2-3-8-7(10)5-6-9(14)11(8)15/h2-6,9,11,14-15H,1H3,(H,13,16)
Formula Weight 235.0845
Chemical Name or Material Carbaryl-5,6-dihydro-5,6-dihydroxy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.