missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro Thymine, TRC

Product Code. 30134189
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134189 1 g 1g
30133126 5 g 5g
30008575 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134189 Supplier Toronto Research Chemicals Supplier No. TRCD4494401G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-04-8
Molecular Formula C5 H8 N2 O2
Molecular Weight (g/mol) 128.13
Synonym 2,4(1H,3H)-Pyrimidinedione, dihydro-5-methyl-, Hydrouracil, 5-methyl- (6CI,8CI), Dihydro-5-methyl-2,4(1H,3H)-pyrimidinedione, 5,6-Dihydro-5-methyluracil, 5,6-Dihydrothymine, 5-Methyl-5,6-dihydrouracil, 5-Methyldihydropyrimidine-2,4(1H,3H)-dione, Dihydrothymine, NSC 44131
IUPAC Name 5-methyl-1,3-diazinane-2,4-dione
SMILES CC1CNC(=O)NC1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H8N2O2/c1-3-2-6-5(9)7-4(3)8/h3H,2H2,1H3,(H2,6,7,8,9)
Formula Weight 128.0586
Chemical Name or Material 5,6-Dihydrothymine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.