missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-5-methyluracil, 98+%

Product Code. 11381467
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381467 5 g 5g
11391467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381467 Supplier Thermo Scientific Alfa Aesar Supplier No. L01996.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 696-04-8 | C5H8N2O2 | 128.13 g/mol

5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-04-8
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00023159
InChI Key NBAKTGXDIBVZOO-UHFFFAOYNA-N
Synonym dihydrothymine, 5,6-dihydro-5-methyluracil, 5,6-dihydrothymine, 5-methyl-5,6-dihydrouracil, hydrouracil, 5-methyl, 5-methyl-hydrouracil, 5-methyldihydropyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, dihydro-5-methyl, 5,6-dihydro-5-methyl-2,4-dioxypyrimidine, 5,6-dihydro-2,4-dihydroxy-5-methylpyrimidine
PubChem CID 93556
ChEBI CHEBI:27468
IUPAC Name 5-methyl-1,3-diazinane-2,4-dione
SMILES CC1CNC(=O)NC1=O

Specifications

Melting Point 263°C to 265°C
Quantity 5 g
Beilstein 81983
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 128.13
Percent Purity ≥98%
Chemical Name or Material 5,6-Dihydro-5-methyluracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.