missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-2H-pyran-2-one, 90%

Product Code. 10041374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041374 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10041374 Supplier Thermo Scientific Acros Supplier No. 426640250
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3393-45-1 | C5H6O2 | 98.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3393-45-1
Density 1.1390g/mL
Boiling Point 103.0°C (10.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C5H6O2
MDL Number MFCD00010439
Quantity 25 g
Specific Gravity 1.139
Synonym 5,6-dihydro-2h-pyran-2-one, 2h-pyran-2-one, 5,6-dihydro, 5,6-dihydropyran-2-one, 5,6-dihydro-pyran-2-one, chembl55078, 5,6-dihydro-2-pyranone, acmc-1ck70, 2-pentenoic acid, 5-hydroxy-, .delta.-lactone, 5,6-dihydro-2h-pyran-2-one, technical grade
InChI Key VREFDQNWPNDZEX-UHFFFAOYSA-N
SMILES O=C1CC=CCO1
IUPAC Name 2,3-dihydropyran-6-one
Molecular Weight (g/mol) 98.10
PubChem CID 520660
Formula Weight 98.1
Percent Purity 90%
Chemical Name or Material 5,6-Dihydro-2H-pyran-2-one
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5,6-Dihydro-2H-pyran-2-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.