missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloropurine-1-Beta-D-ribofuanosyl-H-benzimidazole, TRC

Product Code. 30385034
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
Product Code. Quantity
30385034 1 g
30371577 10 mg
30368930 250 mg
3 options
This item is not returnable. View return policy

Product Code. 30385034

Brand: Toronto Research Chemicals TRCD4358201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-85-0
Formula Weight 318.0174
InChI Formula InChI=1S/C12H12Cl2N2O4/c13-5-1-7-8(2-6(5)14)16(4-15-7)12-11(19)10(18)9(3-17)20-12/h1-2,4,9-12,17-19H,3H2/t9-,10-,11-,12-/m1/s1
IUPAC Name (2R,3R,4S,5R)-2-(5,6-dichlorobenzimidazol-1-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Formula C12 H12 Cl2 N2 O4
Molecular Weight (g/mol) 319.14
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3cc(Cl)c(Cl)cc23
Synonym 5,6-Dichloropurine-1-B-D-Ribofuanosyl-H-Benzimidazole
Chemical Name or Material 5,6-Dichloropurine-1-beta-D-ribofuanosyl-H-benzimidazole

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.