missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloronicotinic acid, 98%, Thermo Scientific™

Product Code. 10270553 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10270553 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10270553 Supplier Thermo Scientific Acros Supplier No. 378790050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 41667-95-2
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H3Cl2NO2
MDL Number MFCD00075181
Quantity 5g
Synonym 5,6-dichloronicotinic acid, 5,6-dichloro nicotinic acid, 5,6-dichloro-3-pyridinecarboxylic acid, 5,6-dichloronicotinicacid, 2,3-dichloro-5-carboxypyridine, 3-pyridinecarboxylic acid, 5,6-dichloro, 5,6-dichloro-nicotinic acid, pubchem12968, acmc-209jl4, 5,6 dichloronicotinic acid
InChI Key RNRLTTNKVLFZJS-UHFFFAOYSA-N
SMILES C1=C(C=NC(=C1Cl)Cl)C(=O)O
IUPAC Name 5,6-dichloropyridine-3-carboxylic acid
Molecular Weight (g/mol) 191.995
PubChem CID 247987
Formula Weight 192
Percent Purity ≥97.5%
Chemical Name or Material 5, 6-Dichloronicotinic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.