missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichlorobenzimidazole riboside, 98%

Product Code. 15456619
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456619 250 mg 250mg
15466619 50 mg 50mg
15446619 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15456619 Supplier Thermo Scientific Alfa Aesar Supplier No. J64870.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53-85-0 | C12H12Cl2N2O4 | 319.138 g/mol

Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.5,6-Dichlorobenzimidazole riboside is used as an inhibitor of RNA Polymerase II. It is nucleoside analogue and used in research to study the mechanisms of cellular regulation. Further, it inhibits some protein kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibits casein kinase II and blocks RNA synthesis. An adenosine analog.5,6-Dichlorobenzimidazole riboside is used as an inhibitor of RNA Polymerase II. It is nucleoside analogue and used in research to study the mechanisms of cellular regulation. Further, it inhibits some protein kinases.

Solubility
Soluble in DMSO to 75mMSoluble in dimethyl sulfoxide, hot ethanol, dimethyl formamide, (1:2) dimethyl formamide:Phosphate buffer saline, water, methanol and pyridine. Slightly soluble in aqueous buffers.

Notes
Incompatible with heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-85-0
Molecular Formula C12H12Cl2N2O4
Molecular Weight (g/mol) 319.138
MDL Number MFCD00036785
InChI Key XHSQDZXAVJRBMX-DDHJBXDOSA-N
Synonym 5,6-dichloro-1-beta-d-ribofuranosylbenzimidazole, dichlororibofuranosylbenzimidazole, drb, 5,6-dichlorobenzimidazole riboside, 5,6-dichlorobenzimidazole 1-beta-d-ribofuranoside, 5,6-dichloro-1-.beta.-d-ribofuranosylbenzimidazole, benzimidazole, 5,6-dichloro-1-beta-d-ribofuranosyl, 5,6-dichloro-1-beta-d-ribofuranosyl-1h-benzimidazole, 2r,3r,4s,5r-2-5,6-dichloro-1,3-benzodiazol-1-yl-5-hydroxymethyl oxolane-3,4-diol, 2r,3r,4s,5r-2-5,6-dichloro-1h-1,3-benzodiazol-1-yl-5-hydroxymethyl oxolane-3,4-diol
PubChem CID 5894
IUPAC Name (2R,3R,4S,5R)-2-(5,6-dichlorobenzimidazol-1-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=C2C(=CC(=C1Cl)Cl)N(C=N2)C3C(C(C(O3)CO)O)O

Specifications

Color White
Quantity 250 mg
Beilstein 39123
Solubility Information Soluble in DMSO to 75mMSoluble in dimethyl sulfoxide,hot ethanol,dimethyl formamide,(1:2) dimethyl formamide:Phosphate buffer saline,water,methanol and pyridine; Slightly soluble in aqueous buffers.
Formula Weight 319.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5,6-Dichlorobenzimidazole riboside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.