missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5(6)-CFDA

Product Code. 16488896
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488896 100 mg 100mg
16414677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488896 Supplier MedChemExpress Supplier No. HYD0722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5(6)-CFDA (5-(6)-Carboxyfluorescein diacetate) is a cell-permeant, amine-reactive green fluorescent probe used to stain cells for analysis in applications. (λex: 492 nm; λem: 517 nm).

Specifications

Content And Storage -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : 125 mg/mL (271.51 mM; Need ultrasonic)
Target HBV
Molecular Weight (g/mol) 460.39 g/mol
CAS 124387-19-5
Synonym 5-(6)-Carboxyfluorescein diacetate ∣ CFDA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.