missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid, Mikromol

Product Code. 30461615
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461615 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461615 Supplier Mikromol Supplier No. MM3779.080025
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid
CAS 4242-18-6
Analyte or Component Names 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C11 H12 O2
Certifications/Compliance ISO 17025
Synonym 1-Naphthalenecarboxylic acid, 5,6,7,8-tetrahydro-, 1-Naphthoic acid, 5,6,7,8-tetrahydro- (6CI,7CI,8CI), 5,6,7,8-Tetrahydro-1-naphthalenecarboxylic acid, 5,6,7,8-Tetrahydro-1-naphthoic acid, 5,6,7,8-Tetrahydronaphthalen-1-carboxylic acid, NSC 44874, 5,6,7,8-Tetrahydronaphthalene-1-carboxylic Acid
InChI Formula InChI=1S/C11H12O2/c12-11(13)10-7-3-5-8-4-1-2-6-9(8)10/h3,5,7H,1-2,4,6H2,(H,12,13)
Solution Type Neat
SMILES OC(=O)c1cccc2CCCCc12
IUPAC Name 5,6,7,8-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 176.21
Quantity 25 mg
Formula Weight 176.0837 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.