missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-1-naphthylamine, 98%, Thermo Scientific™

Product Code. 10294410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10294410 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10294410 Supplier Thermo Scientific Acros Supplier No. 104650250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2217-41-6
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00001736
InChI Key SODWJACROGQSMM-UHFFFAOYSA-N
Synonym 5,6,7,8-tetrahydro-1-naphthylamine, 5-aminotetralin, 1-amino-5,6,7,8-tetrahydronaphthalene, 5-tetralylamine, 1-naphthalenamine, 5,6,7,8-tetrahydro, 1-naphthylamine, 5,6,7,8-tetrahydro, 5-amino-1,2,3,4-tetrahydronaphthalene, tetralin-5-amine, alpha-tetrahydronaphthylamine, 5,6,7,8-tetrahydro-1-naphthalenamine
PubChem CID 16671
IUPAC Name 5,6,7,8-tetrahydronaphthalen-1-amine
SMILES NC1=CC=CC2=C1CCCC2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38.0°C
Color Yellow to Brown
Density 1.0560g/mL
Boiling Point 275.0°C to 277.0°C (713.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 12, 1197
Specific Gravity 1.056
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in organic solvents (20°C)
Formula Weight 147.22
Percent Purity 98%
Grade Technical
Physical Form Liquid
Chemical Name or Material 5,6,7,8-Tetrahydro-1-naphthylamine, tech., 90%
Show More Show Less
compliance-icons
Product Identifier
  • 5,6,7,8-Tetrahydro-1-naphthylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.