missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetradehydro Risperidone, TRC

Product Code. 30413910
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413910 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413910 Supplier Toronto Research Chemicals Supplier No. TRCT291650100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,6,7,8-Tetradehydro Risperidone
CAS 108855-18-1
Molecular Formula C23 H23 F N4 O2
Quantity 100 mg
Synonym 4H-Pyrido[1,2-a]pyrimidin-4-one','3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-2-methyl-, 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 1,2-Benzisoxazole','4H-pyrido[1,2-a]pyrimidin-4-one deriv.
InChI Formula InChI=1S/C23H23FN4O2/c1-15-18(23(29)28-10-3-2-4-21(28)25-15)9-13-27-11-7-16(8-12-27)22-19-6-5-17(24)14-20(19)30-26-22/h2-6,10,14,16H,7-9,11-13H2,1H3
SMILES CC1=C(CCN2CCC(CC2)c3noc4cc(F)ccc34)C(=O)N5C=CC=CC5=N1
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methylpyrido[1,2-a]pyrimidin-4-one
Molecular Weight (g/mol) 406.45
Formula Weight 406.1805
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.