missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,7a-Tetrahydrothieno[3,2-c]pyridinone Hydrochloride, TRC

Product Code. 30412240
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30412240 5 g 5g
30412239 1 g -
2 options
This item is not returnable. View return policy

Product Code. 30412240

Brand: Toronto Research Chemicals TRCT2957805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115473-15-9
Formula Weight 191.0172
InChI Formula InChI=1S/C7H9NOS.ClH/c9-7-3-5-4-8-2-1-6(5)10-7;/h3,6,8H,1-2,4H2;1H
IUPAC Name 5,6,7,7a-tetrahydro-4H-thieno[3,2-c]pyridin-2-one;hydrochloride
Molecular Formula C7 H9 N O S . Cl H
Molecular Weight (g/mol) 191.68
SMILES Cl.O=C1SC2CCNCC2=C1
Synonym Prasugrel Impurity 7
Chemical Name or Material 5,6,7,7a-Tetrahydro-thieno[3,2-c]pyridinone Hydrochloride

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.