missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Methylenedisalicylic acid, 95%

Product Code. 10536641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10536641 5 g Glass bottle 5g
10205771 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10536641 Supplier Thermo Scientific Acros Supplier No. 351730050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-25-8 | C15H12O6 | 288.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-25-8
Molecular Formula C15H12O6
Molecular Weight (g/mol) 288.26
MDL Number MFCD00016506
InChI Key JWQFKVGACKJIAV-UHFFFAOYSA-N
Synonym 5,5'-methylenedisalicylic acid, 5,5'-methylenebis 2-hydroxybenzoic acid, methylenebis salicylic acid, 5,5-methylenebis salicylic acid, unii-2kf4fvv76n, 5,5-methylenedisalicylic acid, 5-3-carboxy-4-hydroxybenzyl salicylic acid, 3,3'-dicarboxy-4,4'-dihydroxydiphenylmethane, 4,4'-dihydroxy-3,3'-dicarboxydiphenylmethane, 2kf4fvv76n
PubChem CID 67145
IUPAC Name 5-[(3-carboxy-4-hydroxyphenyl)methyl]-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1CC2=CC(=C(C=C2)O)C(=O)O)C(=O)O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 235°C to 242°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH2[C6H3(OH)COOH]2
Quantity 5 g
Merck Index 15, 6136
Solubility Information Solubility in water: insoluble in water
Formula Weight 288.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5, 5'-Methylenedisalicylic acid, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.